Assay Details
| Gene Symbol: SSNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015094098.2 | XP_014949584.2 | 1 - 2 | 132 | 68 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

SSNA1| Gene Symbol: SSNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015094098.2 | XP_014949584.2 | 1 - 2 | 132 | 68 | |