Assay Details
Gene Symbol: LOC101108197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027965595.1 | XP_027821396.1 | 3 - 4 | 366 | 89 | |
XM_027965596.1 | XP_027821397.1 | 3 - 4 | 426 | 89 | ||
XM_027965597.1 | XP_027821398.1 | 3 - 4 | 402 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
