Assay Details
| Gene Symbol: PDLIM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962546.1 | XP_027818347.1 | 1 - 2 | 178 | 60 | |
| XM_027962547.1 | XP_027818348.1 | 1 - 2 | 178 | 60 | ||
| XM_027962548.1 | XP_027818349.1 | 1 - 2 | 178 | 60 | ||
More Information
Set Membership: |
|

PDLIM3
Genomic Map