Assay Details
Gene Symbol: CARD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964198.1 | XP_027819999.1 | 2 - 3 | 216 | 93 | |
XM_027964199.1 | XP_027820000.1 | 2 - 3 | 568 | 93 | ||
XM_027964200.1 | XP_027820001.1 | 2 - 3 | 216 | 93 | ||
XM_027964201.1 | XP_027820002.1 | 2 - 3 | 567 | 93 | ||
XM_027964202.1 | XP_027820003.1 | 2 - 3 | 216 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
