Assay Details
| Gene Symbol: MARCKSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015093618.2 | XP_014949104.2 | 1 - 2 | 325 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MARCKSL1| Gene Symbol: MARCKSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015093618.2 | XP_014949104.2 | 1 - 2 | 325 | 93 | |
Set Membership: |
|