Assay Details
| Gene Symbol: SYT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979417.1 | XP_027835218.1 | 2 - 3 | 500 | 76 | |
| XM_027979418.1 | XP_027835219.1 | 2 - 3 | 500 | 76 | ||
More Information
Set Membership: |
|

SYT9
Genomic Map