Assay Details
| Gene Symbol: AP3S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015096778.2 | XP_014952264.1 | 1 - 2 | 200 | 64 | |
| XM_015096779.2 | XP_014952265.1 | 1 - 2 | 180 | 64 | ||
| XM_027971459.1 | XP_027827260.1 | 1 - 2 | 173 | 64 | ||
More Information
Set Membership: |
|

AP3S1
Genomic Map