Assay Details
| Gene Symbol: POLD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968729.1 | XP_027824530.1 | 11 - 12 | 1510 | 61 | |
| XM_027968730.1 | XP_027824531.1 | 11 - 12 | 1712 | 61 | ||
| XM_027968731.1 | XP_027824532.1 | 10 - 11 | 1420 | 61 | ||
More Information
Set Membership: |
|

POLD2
Genomic Map