Assay Details
Gene Symbol: GFY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977592.1 | XP_027833393.1 | 3 - 4 | 1349 | 88 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GFY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027977592.1 | XP_027833393.1 | 3 - 4 | 1349 | 88 |