Assay Details
Gene Symbol: B3GALT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958660.1 | XP_027814461.1 | 2 | 665 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
