Assay Details
| Gene Symbol: CACFD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027966146.1 | XP_027821947.1 | 1 - 2 | 190 | 85 | |
| XM_027966147.1 | XP_027821948.1 | 1 - 2 | 180 | 85 | ||
More Information
Set Membership: |
|

CACFD1
Genomic Map