Assay Details
| Gene Symbol: LOC101114342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006170.4 | XP_004006219.1 | 3 - 4 | 624 | 91 | |
| XM_012173857.3 | XP_012029247.1 | 4 - 5 | 668 | 91 | ||
More Information
Set Membership: |
|

LOC101114342
Genomic Map