Assay Details
Gene Symbol: LOC101117600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013284.4 | XP_004013333.1 | 2 - 3 | 365 | 85 | |
XM_027975008.1 | XP_027830809.1 | 2 - 3 | 366 | 85 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
