Assay Details
| Gene Symbol: ALKBH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017618.3 | XP_004017667.3 | 3 - 4 | 961 | 75 | |
| XM_012097947.3 | XP_011953337.2 | 3 - 4 | 718 | 75 | ||
| XM_012097948.3 | XP_011953338.2 | 3 - 4 | 730 | 75 | ||
More Information
Set Membership: |
|

ALKBH2
Genomic Map