Assay Details
| Gene Symbol: WASHC2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021567.3 | XP_004021616.3 | 14 - 15 | 1300 | 54 | |
| XM_027962407.1 | XP_027818208.1 | 14 - 15 | 1297 | 54 | ||
More Information
Set Membership: |
|

WASHC2C
Genomic Map