Assay Details
| Gene Symbol: WASHC2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021567.3 | XP_004021616.3 | 8 - 9 | 796 | 81 | |
| XM_027962407.1 | XP_027818208.1 | 8 - 9 | 793 | 81 | ||
More Information
Set Membership: |
|

WASHC2C
Genomic Map