Assay Details
| Gene Symbol: PIGU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012189096.3 | XP_012044486.2 | 10 - 11 | 1084 | 63 | |
| XM_027977104.1 | XP_027832905.1 | 9 - 10 | 1005 | 63 | ||
More Information
Set Membership: |
|

PIGU
Genomic Map