Assay Details
Gene Symbol: LOC101115706 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969603.1 | XP_027825404.1 | 2 - 3 | 341 | 86 | |
XM_027969604.1 | XP_027825405.1 | 1 - 2 | 87 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027969604.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
