Assay Details
Gene Symbol: KLHL25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004017732.3 | XP_004017781.2 | 2 - 3 | 2096 | 105 | |
XM_012098433.2 | XP_011953823.1 | 2 - 3 | 2099 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
