Assay Details
Gene Symbol: MSMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004021566.4 | XP_004021615.1 | 3 - 4 | 312 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
