Assay Details
| Gene Symbol: RGS20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011680.3 | XP_004011729.2 | 4 - 5 | 611 | 75 | |
| XM_027972956.1 | XP_027828757.1 | 5 - 6 | 1070 | 75 | ||
| XM_027972957.1 | XP_027828758.1 | 5 - 6 | 1052 | 75 | ||
More Information
Set Membership: |
|

RGS20
Genomic Map