Assay Details
Gene Symbol: SLC30A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011988.4 | XP_004012037.2 | 4 - 5 | 584 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
