Assay Details
| Gene Symbol: HPS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956674.1 | XP_027812475.1 | 5 - 6 | 956 | 58 | |
| XM_027956675.1 | XP_027812476.1 | 5 - 6 | 931 | 58 | ||
| XM_027956676.1 | XP_027812477.1 | 5 - 6 | 956 | 58 | ||
| XM_027956677.1 | XP_027812478.1 | 5 - 6 | 956 | 58 | ||
| XM_027956678.1 | XP_027812479.1 | 5 - 6 | 956 | 58 | ||
More Information
Set Membership: |
|

HPS4
Genomic Map