Assay Details
| Gene Symbol: GALE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965533.1 | XP_027821334.1 | 3 - 4 | 409 | 67 | |
| XM_027965534.1 | XP_027821335.1 | 3 - 4 | 384 | 67 | ||
More Information
Set Membership: |
|

GALE
Genomic Map