Assay Details
Gene Symbol: LOC101121385 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015092152.2 | XP_014947638.2 | 5 - 6 | 514 | 95 | |
XM_027964822.1 | XP_027820623.1 | 5 - 6 | 508 | 95 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
