Assay Details
Gene Symbol: RETREG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004940.4 | XP_004004989.2 | 4 - 5 | 648 | 66 | |
XM_027965206.1 | XP_027821007.1 | 3 - 4 | 124 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027965206.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
