Assay Details
| Gene Symbol: TECRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004009860.4 | XP_004009909.1 | 6 - 7 | 709 | 83 | |
| XM_027971027.1 | XP_027826828.1 | 6 - 7 | 710 | 83 | ||
More Information
Set Membership: |
|

TECRL
Genomic Map