Assay Details
| Gene Symbol: MLST8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961182.1 | XP_027816983.1 | 6 - 7 | 719 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MLST8| Gene Symbol: MLST8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961182.1 | XP_027816983.1 | 6 - 7 | 719 | 58 | |
Set Membership: |
|