Assay Details
Gene Symbol: LOC114108784 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027956248.1 | XP_027812049.1 | 3 - 4 | 2124 | 163 |
Gene Symbol: DTX3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027956374.1 | XP_027812175.1 | 3 - 4 | 1955 | 163 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
