Assay Details
| Gene Symbol: ANAPC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017223.4 | XP_004017272.1 | 2 - 3 | 255 | 101 | |
| XM_012097422.3 | XP_011952812.1 | 2 - 3 | 398 | 101 | ||
| XM_012097423.3 | XP_011952813.1 | 2 - 3 | 212 | 101 | ||
| XM_027956255.1 | XP_027812056.1 | 2 - 3 | 212 | 101 | ||
More Information
Set Membership: |
|

ANAPC10
Genomic Map