Assay Details
Gene Symbol: LOC101104287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015103582.2 | XP_014959068.1 | 5 - 6 | 580 | 121 | |
XM_027960758.1 | XP_027816559.1 | 5 - 6 | 682 | 121 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
