Assay Details
Gene Symbol: GAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959592.1 | XP_027815393.1 | 4 - 5 | 381 | 53 | |
GenBank mRNA | EF192581.1 | - | 3 - 4 | 230 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() |
