Assay Details
| Gene Symbol: LOC443459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012180067.3 | XP_012035457.2 | 17 - 18 | 2790 | 109 | |
| XM_012180068.3 | XP_012035458.2 | 17 - 18 | 2779 | 109 | ||
| XM_012180069.3 | XP_012035459.2 | 17 - 18 | 2882 | 109 | ||
| GenBank mRNA | U35657.1 | - | 8 - 9 | 1044 | 109 | |
More Information
Set Membership: |
|

LOC443459
Genomic Map