Assay Details
| Gene Symbol: LOC443459 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012180067.3 | XP_012035457.2 | 7 - 8 | 960 | 99 | |
| XM_012180069.3 | XP_012035459.2 | 7 - 8 | 1052 | 99 | ||
More Information
Set Membership: |
|

LOC443459
Genomic Map