Assay Details
| Gene Symbol: LOC101104808 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004811.4 | XP_004004860.2 | 4 - 5 | 413 | 83 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101104808| Gene Symbol: LOC101104808 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004004811.4 | XP_004004860.2 | 4 - 5 | 413 | 83 | |