Assay Details
| Gene Symbol: EMG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006919.4 | XP_004006968.3 | 1 - 2 | 346 | 77 | |
| XM_027967923.1 | XP_027823724.1 | 1 - 2 | 354 | 77 | ||
| XM_027967924.1 | XP_027823725.1 | 1 - 2 | 374 | 77 | ||
| XM_027967925.1 | XP_027823726.1 | 1 - 2 | 370 | 77 | ||
| XM_027967926.1 | XP_027823727.1 | 1 - 2 | 364 | 77 | ||
More Information
Set Membership: |
|

EMG1
Genomic Map