Assay Details
Gene Symbol: LOC101113487 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004007023.3 | XP_004007072.1 | 8 - 9 | 982 | 99 | |
XM_012175713.2 | XP_012031103.1 | 8 - 9 | 979 | 99 | ||
XM_012175714.2 | XP_012031104.1 | 8 - 9 | 982 | 99 | ||
XM_012175715.3 | XP_012031105.1 | 8 - 9 | 975 | 99 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
