Assay Details
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962427.1 | XP_027818228.1 | 2 - 3 | 1579 | 86 | |
XM_027962428.1 | XP_027818229.1 | 2 - 3 | 1591 | 86 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027962427.1 | XP_027818228.1 | 2 - 3 | 1579 | 86 | |
XM_027962428.1 | XP_027818229.1 | 2 - 3 | 1591 | 86 |