Assay Details
| Gene Symbol: LOC101110605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012186735.3 | XP_012042125.3 | 2 - 3 | 274 | 72 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101110605| Gene Symbol: LOC101110605 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012186735.3 | XP_012042125.3 | 2 - 3 | 274 | 72 | |