Assay Details
| Gene Symbol: CAPS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969798.1 | XP_027825599.1 | 2 - 3 | 1665 | 100 | |
| XM_027969799.1 | XP_027825600.1 | 2 - 3 | 1546 | 100 | ||
More Information
Set Membership: |
|

CAPS
Genomic Map