Assay Details
| Gene Symbol: TRMT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015092136.2 | XP_014947622.1 | 4 - 5 | 474 | 79 | |
| XM_015092137.2 | XP_014947623.1 | 4 - 5 | 498 | 79 | ||
| XM_027972341.1 | XP_027828142.1 | 4 - 5 | 443 | 79 | ||
More Information
Set Membership: |
|

TRMT13
Genomic Map