Assay Details
Gene Symbol: PTP4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027972826.1 | XP_027828627.1 | 3 - 4 | 1160 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
