Assay Details
| Gene Symbol: SMPD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011902.4 | XP_004011951.2 | 2 - 3 | 1061 | 68 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

SMPD5| Gene Symbol: SMPD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011902.4 | XP_004011951.2 | 2 - 3 | 1061 | 68 | |