Assay Details
Gene Symbol: MINDY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004014242.4 | XP_004014291.1 | 8 - 9 | 946 | 79 | |
XM_012188433.3 | XP_012043823.1 | 8 - 9 | 672 | 79 | ||
XM_027960112.1 | XP_027815913.1 | 7 - 8 | 894 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
