Assay Details
Gene Symbol: LOC101118315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959706.1 | XP_027815507.1 | 1 - 2 | 122 | 102 | |
XM_027959707.1 | XP_027815508.1 | 1 - 2 | 137 | 102 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
