Assay Details
| Gene Symbol: DDX50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021419.4 | XP_004021468.1 | 9 - 10 | 1523 | 67 | |
| XM_012105204.3 | XP_011960594.1 | 10 - 11 | 1518 | 67 | ||
| XM_027962173.1 | XP_027817974.1 | 9 - 10 | 1528 | 67 | ||
More Information
Set Membership: |
|

DDX50
Genomic Map