Assay Details
| Gene Symbol: STOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962614.1 | XP_027818415.1 | 1 - 2 | 3166 | 85 | |
| XM_027962615.1 | XP_027818416.1 | 1 - 2 | 3166 | 85 | ||
| XM_027962616.1 | XP_027818417.1 | 1 - 2 | 3166 | 85 | ||
More Information
Set Membership: |
|

STOX2
Genomic Map