Assay Details
| Gene Symbol: MAPK12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027968252.1 | XP_027824053.1 | 10 - 11 | 1047 | 66 | |
| XM_027968254.1 | XP_027824055.1 | 10 - 11 | 997 | 66 | ||
| XM_027968255.1 | XP_027824056.1 | 9 - 10 | 790 | 66 | ||
More Information
Set Membership: |
|

MAPK12
Genomic Map