Assay Details
| Gene Symbol: CES3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012189895.3 | XP_012045285.2 | 11 - 12 | 1506 | 59 | |
| XM_015100471.2 | XP_014955957.2 | 11 - 12 | 1471 | 59 | ||
More Information
Set Membership: |
|

CES3
Genomic Map