Assay Details
| Gene Symbol: BICD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015093028.2 | XP_014948514.2 | 6 - 7 | 2293 | 63 | |
| XM_027964209.1 | XP_027820010.1 | 6 - 7 | 2293 | 63 | ||
| XM_027964210.1 | XP_027820011.1 | 5 - 6 | 3307 | 63 | ||
More Information
Set Membership: |
|

BICD2
Genomic Map